ENA964482299 Enamine

propyl 2-hydroxybut-3-enoate

  • MDL number MFCD31439545
  • Cas Number : 1403229-32-2
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-717396

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-717396
Availability details for
Quantity
  • Availability
  • Add to Cart