ENA964485613 Enamine

3-cyclopentylpiperidine

  • MDL number MFCD20547689
  • Cas Number : 794533-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-722134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-722134
Availability details for
Quantity
  • Availability
  • Add to Cart