ENA964486844 Enamine

sodium 6-methylpyridazine-3-sulfinate

  • MDL number MFCD31441722
  • Cas Number : 2137835-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-723559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-723559
Availability details for
Quantity
  • Availability
  • Add to Cart