ENA964487275 Enamine

sodium 1-methyl-1H-1,2,3-triazole-4-sulfinate

  • MDL number MFCD31442189
  • Cas Number : 2137669-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-724054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-724054
Availability details for
Quantity
  • Availability
  • Add to Cart