ENA964488765 Enamine

sodium 1-[(tert-butoxy)carbonyl]pyrrolidine-3-sulfinate

  • MDL number MFCD31442776
  • Cas Number : 2138279-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-724672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-724672
Availability details for
Quantity
  • Availability
  • Add to Cart