ENA964489050 Enamine

1-(trifluoromethyl)cyclobutane-1-carbonitrile

  • MDL number MFCD31442988
  • Cas Number : 1447949-37-2
  • Molecular Weight : 149.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-725010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-725010
Availability details for
Quantity
  • Availability
  • Add to Cart