ENA964506820 Enamine

2-(2-ethoxyethoxy)ethane-1-sulfonyl chloride

  • MDL number MFCD19669371
  • Cas Number : 1343089-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-738729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-738729
Availability details for
Quantity
  • Availability
  • Add to Cart