ENA964511260 Enamine

methyl 5-amino-3-tert-butyl-1,2-thiazole-4-carboxylate

  • MDL number MFCD31453645
  • Cas Number : 2138226-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-742108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-742108
Availability details for
Quantity
  • Availability
  • Add to Cart