ENA964513775 Enamine

3-(4-methoxyphenyl)prop-2-ynoic acid

  • MDL number MFCD00181523
  • Cas Number : 2227-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-743493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-743493
Availability details for
Quantity
  • Availability
  • Add to Cart