ENA964518171 Enamine

(1R,2S)-2-methylcyclopropane-1-carboxylic acid

  • MDL number MFCD12031257
  • Cas Number : 29667-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66625
Availability details for
Quantity
  • Availability
  • Add to Cart