ENAH03855CC1 Enamine

2-amino-1-[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethan-1-ol

  • MDL number MFCD31421192
  • Cas Number : 2089622-54-6
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-366335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366335
Availability details for
Quantity
  • Availability
  • Add to Cart