ENAH03855CED Enamine

(1-benzyl-3-methylpiperidin-4-yl)methanamine

  • MDL number MFCD31421509
  • Cas Number : 2138430-68-7
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-622275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-622275
Availability details for
Quantity
  • Availability
  • Add to Cart