ENAH03855D40 Enamine

2-bromoprop-2-enenitrile

  • MDL number MFCD00040191
  • Cas Number : 920-34-3
  • Molecular Weight : 131.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Flash Point 42
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-140716

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-140716
pictograms GHS02,GHS05,GHS07
hstatements H226,H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart