ENAH03855DD9 Enamine

5-(propan-2-yl)pyridin-3-amine

  • MDL number MFCD18256499
  • Cas Number : 1256821-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203422
Availability details for
Quantity
  • Availability
  • Add to Cart