ENAH03856026 Enamine

methyl 2-(2-hydroxyethyl)-1-benzofuran-6-carboxylate

  • MDL number MFCD31421470
  • Cas Number : 249758-23-4
  • Molecular Weight : 220.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-396185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-396185
Availability details for
Quantity
  • Availability
  • Add to Cart