ENAH03856248 Enamine

methyl 3-amino-4-bromo-5-tert-butylthiophene-2-carboxylate

  • MDL number MFCD31449517
  • Cas Number : 2137591-79-6
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-736111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-736111
Availability details for
Quantity
  • Availability
  • Add to Cart