ENAH038563C0 Enamine

1-phenyl-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD28656978
  • Cas Number : 1706432-02-1
  • Molecular Weight : 279.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-736531

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-736531
pictograms GHS07
hstatements H303,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart