ENAH038564EC Enamine

methyl 2-bromo-3-nitropyridine-4-carboxylate

  • MDL number MFCD17214973
  • Cas Number : 1379365-85-1
  • Molecular Weight : 261.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-746201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-746201
Availability details for
Quantity
  • Availability
  • Add to Cart