ENAH038566A5 Enamine

methyl 1-methoxycycloheptane-1-carboxylate

  • MDL number MFCD31457003
  • Cas Number : 2137929-87-2
  • Molecular Weight : 186.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-747135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-747135
Availability details for
Quantity
  • Availability
  • Add to Cart