ENAH03856720 Enamine

lithium(1+) ion 5-[(tert-butoxy)carbonyl]-4H,5H,6H,7H-[1,3]thiazolo[4,5-c]pyridine-2-carboxylate

  • MDL number MFCD31457161
  • Cas Number : 2137804-12-5
  • Molecular Weight : 290.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-760933

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-760933
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart