ENAH03856738 Enamine

4-(4,5-dihydro-1H-imidazol-2-yl)oxan-4-amine dihydrochloride

  • MDL number MFCD31457190
  • Molecular Weight : 242.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-761254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-761254
Availability details for
Quantity
  • Availability
  • Add to Cart