ENAH03856846 Enamine

4-[ethyl({[(9H-fluoren-9-yl)methoxy]carbonyl})amino]butanoic acid

  • MDL number MFCD31457130
  • Cas Number : 1446478-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-755406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-755406
Availability details for
Quantity
  • Availability
  • Add to Cart