ENAH03856869 Enamine

1-[(2-amino-1,3-thiazol-5-yl)methyl]-3-methylpiperidin-4-amine

  • MDL number MFCD31457156
  • Cas Number : 1881414-78-3
  • Molecular Weight : 226.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-760920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-760920
Availability details for
Quantity
  • Availability
  • Add to Cart