ENAH0385693B Enamine

(3R)-3-{[(tert-butoxy)carbonyl]amino}-5-methylhexanoic acid

  • MDL number MFCD01076256
  • Cas Number : 146398-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-761700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-761700
Availability details for
Quantity
  • Availability
  • Add to Cart