ENAH03856963 Enamine

2-(2,2-difluoroethoxy)-5-methylaniline hydrochloride

  • MDL number MFCD15142914
  • Cas Number : 1431968-06-7
  • Molecular Weight : 223.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-762605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-762605
Availability details for
Quantity
  • Availability
  • Add to Cart