ENAH038569DC Enamine

1-(1-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}ethyl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD25371212
  • Cas Number : 1431964-49-6
  • Molecular Weight : 282.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-762731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-762731
Availability details for
Quantity
  • Availability
  • Add to Cart