ENAH03857D3E Enamine

6-(4-hydroxypiperidin-1-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD31459526
  • Cas Number : 1597210-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-767079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-767079
Availability details for
Quantity
  • Availability
  • Add to Cart