ENAH03857D3F Enamine

6-[4-(hydroxymethyl)piperidin-1-yl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD31459527
  • Cas Number : 1603127-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-767080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-767080
Availability details for
Quantity
  • Availability
  • Add to Cart