ENAH03858130 Enamine

2-(1-benzothiophen-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD13619876
  • Cas Number : 1000160-75-7
  • Molecular Weight : 260.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-benzothien-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C14H17BO2S/c1-13(2)14(3,4)17-15(16-13)11-6-5-7-12-10(11)8-9-18-12/h5-9H,1-4H3
Inchi Key KQTHIDLDBILMSE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-768137

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-768137
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart