ENAH038593C7 Enamine

5-chloro-7-fluoro-1-benzofuran-3-carboxylic acid

  • MDL number MFCD29498423
  • Cas Number : 1508825-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-785027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-785027
Availability details for
Quantity
  • Availability
  • Add to Cart