ENAH0385A17E Enamine

4-ethynyl-1,3-thiazole

  • MDL number MFCD18207415
  • Cas Number : 111600-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-787622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-787622
Availability details for
Quantity
  • Availability
  • Add to Cart