ENAH0385A1AD Enamine

3-ethynylpyridazine

  • MDL number MFCD16988473
  • Cas Number : 1017793-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-787669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-787669
Availability details for
Quantity
  • Availability
  • Add to Cart