ENAH0385B7FD Enamine

3-methyl-1-benzothiophen-5-ol

  • MDL number MFCD18451847
  • Cas Number : 3610-07-9
  • Molecular Weight : 164.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-790786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-790786
Availability details for
Quantity
  • Availability
  • Add to Cart