ENAH0385C75F Enamine

tert-butyl N-[2-oxo-2-(pyrimidin-5-yl)ethyl]carbamate

  • MDL number MFCD31468530
  • Cas Number : 2008482-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-797003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-797003
Availability details for
Quantity
  • Availability
  • Add to Cart