ENAH039FDA73 Enamine

5-(4-chlorophenyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD01217348
  • Cas Number : 131172-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153322
Availability details for
Quantity
  • Availability
  • Add to Cart