ENAH039FE832 Enamine

4-cyclobutylpyrimidin-2-amine

  • MDL number MFCD14707182
  • Cas Number : 1215073-20-3
  • Molecular Weight : 149.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1072826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1072826
Availability details for
Quantity
  • Availability
  • Add to Cart