ENAH03A000C1 Enamine

6-aminobicyclo[3.2.0]heptane-3-carboxylic acid hydrochloride

  • MDL number MFCD31559002
  • Cas Number : 2138070-28-5
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1082510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1082510
Availability details for
Quantity
  • Availability
  • Add to Cart