ENAH03A00312 Enamine

3-(4,5-dichloro-3-methyl-1H-pyrazol-1-yl)propan-1-amine hydrochloride

  • MDL number MFCD28962809
  • Cas Number : 1855911-15-7
  • Molecular Weight : 244.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1082560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1082560
Availability details for
Quantity
  • Availability
  • Add to Cart