ENAH03A0039A Enamine

methyl({[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methyl})amine hydrochloride

  • MDL number MFCD28962808
  • Cas Number : 1432030-55-1
  • Molecular Weight : 229.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1082838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1082838
Availability details for
Quantity
  • Availability
  • Add to Cart