ENAH03A013D9 Enamine

(1,4-dimethyl-1H-pyrazol-5-yl)methanol

  • MDL number MFCD28399266
  • Cas Number : 1607024-78-1
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1086954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1086954
Availability details for
Quantity
  • Availability
  • Add to Cart