ENAH03A014AE Enamine

3-[2-chloro-4-(trifluoromethyl)phenoxy]-N-methanesulfonyl-2-nitrobenzamide

  • MDL number MFCD31559506
  • Cas Number : 116493-45-9
  • Molecular Weight : 438.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1212509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1212509
Availability details for
Quantity
  • Availability
  • Add to Cart