ENAH03A01C05 Enamine

3-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD31592622
  • Cas Number : 2138027-90-2
  • Molecular Weight : 377.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-657616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-657616
Availability details for
Quantity
  • Availability
  • Add to Cart