ENAH03A01C67 Enamine

(2R)-2-[benzyl({[(9H-fluoren-9-yl)methoxy]carbonyl})amino]propanoic acid

  • MDL number MFCD31590467
  • Cas Number : 2137135-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-657851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-657851
Availability details for
Quantity
  • Availability
  • Add to Cart