ENAH03A01CEC Enamine

methyl 2-(2-hydroxyquinolin-3-yl)acetate

  • MDL number MFCD18417536
  • Cas Number : 156496-39-8
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-624280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-624280
Availability details for
Quantity
  • Availability
  • Add to Cart