ENAH03A01CF9 Enamine

3-(2-chlorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid

  • MDL number MFCD06656186
  • Cas Number : 678991-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-651008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-651008
Availability details for
Quantity
  • Availability
  • Add to Cart