ENAH03A01CFE Enamine

4-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]-2-methoxybenzoic acid

  • MDL number MFCD31593502
  • Cas Number : 2125396-31-6
  • Molecular Weight : 403.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-651027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-651027
Availability details for
Quantity
  • Availability
  • Add to Cart