ENAH03A01DC0 Enamine

5-chloro-2-iodo-3-nitropyridine

  • MDL number MFCD17014996
  • Cas Number : 899424-60-3
  • Molecular Weight : 284.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-651664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-651664
Availability details for
Quantity
  • Availability
  • Add to Cart