ENAH03A01ED9 Enamine

2-(cyclopentylmethyl)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid

  • MDL number MFCD28389295
  • Cas Number : 1823505-47-0
  • Molecular Weight : 393.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-650149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650149
Availability details for
Quantity
  • Availability
  • Add to Cart