ENAH03A01F67 Enamine

2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(3-methylphenyl)propanoic acid

  • MDL number MFCD30482934
  • Cas Number : 376588-51-1
  • Molecular Weight : 401.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-650407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650407
Availability details for
Quantity
  • Availability
  • Add to Cart