ENAH03A01F7B Enamine

(3S)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[3-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD03428002
  • Cas Number : 507472-20-0
  • Molecular Weight : 455.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-650426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650426
Availability details for
Quantity
  • Availability
  • Add to Cart